Order Entry
Finland
ContactUsLinkComponent
FORTUNA graduated pipette 5mL : 0,05 with suction piston, amber graduation, clear glass, clear calibrated cylinder, with tolerance indication, use of pipetting aid not necessary 1 * 1 items
Catalog # POUL11423204
Supplier:  Poulten & Graf
FORTUNA graduated pipette 5mL : 0,05 with suction piston, amber graduation, clear glass, clear calibrated cylinder, with tolerance indication, use of pipetting aid not necessary 1 * 1 items
Catalog # POUL11423204
Supplier:  Poulten & Graf