Volumetric flasks, class A
100 ml, 14/23, 0,10 ± ml
Catalog # 612-3803
Supplier: VITLAB
Jump to:
Product Details & Documents
PMP (TPX®), transparent, with PP stopper.
- Individually calibrated ring mark
- Imprinted lot number
- Complete with certificate
DIN EN ISO 1042
Certifications: DIN EN ISO 1042
Specifications
- NS:14/23
- Capacity:100 ml
- Seq:03
- Tolerance:0,10 ± ml