Order Entry
Finland
ContactUsLinkComponent
Stand tripod 230x300mm sock,tripods plastic / aluminum, therefore completely corrosion resistant. socket pvc, gray, 30 mm thick, hea vy duty. stand-rod 13 mm aluminum, for threaded connection socket. socket size 230*300 mm rod length 770 positionin 1 * 1 items
Catalog # 229-7493
Supplier:  SEMADENI
Stand tripod 230x300mm sock,tripods plastic / aluminum, therefore completely corrosion resistant. socket pvc, gray, 30 mm thick, hea vy duty. stand-rod 13 mm aluminum, for threaded connection socket. socket size 230*300 mm rod length 770 positionin 1 * 1 items
Catalog # 229-7493
Supplier:  SEMADENI
Frequently Bought Together