Order Entry
Finland
ContactUsLinkComponent
Cover for 200/300 hp ltr industry industrial storage - hard polyethylene (hdpe) gray. affordable and versatile vessel. stackable wit h enough space for hubstaplerarme distance. suitable for liquids only up to about 50% level. 1 * 1 items
Catalog # 215-0259
Supplier:  SEMADENI
Cover for 200/300 hp ltr industry industrial storage - hard polyethylene (hdpe) gray. affordable and versatile vessel. stackable wit h enough space for hubstaplerarme distance. suitable for liquids only up to about 50% level. 1 * 1 items
Catalog # 215-0259
Supplier:  SEMADENI