Order Entry
Finland
Orders LinkContactUsLinkComponent
D-(-)-Mannitol ACS

D-(-)-Mannitol ACS

Glass bottle for solids, 2,5 kg
Catalog # 33342.A4
D-(-)-Mannitol ACS
Catalog # 33342.A4
CAS Number:  69-65-8
D-(-)-Mannitol ACS
D-(-)-Mannitol ACS

Glass bottle for solids, 2,5 kg

CAS Number:  69-65-8
Price
602,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

CAS No.: 69-65-8 Molecular Formula: HOCH2(CHOH)4CH2OH Formula Weight: 182.17 Physical Form: Crystalline Density: 1.489 Appearance: White Odour: Odourless Melting Point: 165-167° Boiling Point: 290-295°/3.5mm Solubility: Soluble in water, alcohol, pyridine, aniline. Insoluble in ether Beilstein Reference: 1721898 Merck Reference: 14,5745 MDL No.: MFCD00064287

Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:14,5745
  • Boiling Pt:290…295 °C (3.5 mmHg)
  • Melting Pt:165…167 °C
  • Density:1.489
  • Storage Temperature:Ambient
  • EINECS:200-711-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg

Specifications