Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Propylene oxide ≥99%

Glass bottle, 100 mL
Catalog # 30765.AE
Flammable
Health
Irritant
Propylene oxide ≥99%
Catalog # 30765.AE
CAS Number:  75-56-9
Propylene oxide ≥99%
Propylene oxide ≥99%

Glass bottle, 100 mL

CAS Number:  75-56-9
Price
57,20 €
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 99+% CAS No.: 75-56-9 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -37°(-35°F) Physical Form: Liquid Density: 0.83 Refractive Index: 1.3660 Appearance: Colourless Odour: Ether-like Melting Point: -112° Boiling Point: 34-35° Beilstein Reference: 79763 Merck Reference: 14,7856 UN No.: 1280 MDL No.: MFCD00005126

Synonyms:  (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide
  • CAS Number:75-56-9
  • Formula:C₃H₆O
  • MDL Number:MFCD00005126
  • Molecular Weight:58.08 g/mol
  • Merck Index:14,7856
  • Boiling Pt:34…35 °C
  • Melting Pt:–112 °C
  • Density:0.83
  • Flash Pt:–37 °C (–35 °F)
  • Storage Temperature:Refrigerator
  • EINECS:200-879-2
  • UN:1280
  • ADR:3,I
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications