Strontium oxide, tech.
Plastic bottle for solids, 1 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 1314-11-0 Molecular Formula: SrO, may also contain Sr(OH)2, SrCO3 Formula Weight: 103.62 Physical Form: -100 Mesh Powder Density: 4.700 Sensitivity: Moisture Sensitive Boiling Point: ca 3000° Solubility: Soluble in fused potassium hydroxide. Insoluble in ether and acetone. Decomposes in water to Sr(OH)2 with evolution of heat Merck Reference: 14,8846 UN No.: 3262 MDL No.: MFCD00049558
In the manufacture of strontium salts, pyrotechnics, and pigments, in greases and soaps, and as a desiccant
- CAS Number:1314-11-0
- Formula:SrO
- MDL Number:MFCD00049558
- Molecular Weight:103.62 g/mol
- Merck Index:14,8846
- Boiling Pt:∼3000 °C
- Melting Pt:2430 °C
- Density:4.700
- Storage Temperature:Ambient
- EINECS:215-219-9
- UN:3262
- ADR:8,II
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg



