Strontium oxide, tech.
Plastic bottle for solids, 250 g
CAS No.: 1314-11-0 Molecular Formula: SrO, may also contain Sr(OH)2, SrCO3 Formula Weight: 103.62 Physical Form: -100 Mesh Powder Density: 4.700 Sensitivity: Moisture Sensitive Boiling Point: ca 3000° Solubility: Soluble in fused potassium hydroxide. Insoluble in ether and acetone. Decomposes in water to Sr(OH)2 with evolution of heat Merck Reference: 14,8846 UN No.: 3262 MDL No.: MFCD00049558
In the manufacture of strontium salts, pyrotechnics, and pigments, in greases and soaps, and as a desiccant
- :1314-11-0
- :SrO
- :MFCD00049558
- :103.62 g/mol
- :14,8846
- :∼3000 °C
- :2430 °C
- :4.700
- :Ambient
- :215-219-9
- :3262
- :8,II
- Pack type:Plastic bottle for solids
- Pk:250 g