Order Entry
Finland
Orders LinkContactUsLinkComponent
3-(2-Hydroxyethyl)indole

3-(2-Hydroxyethyl)indole

Glass bottle for solids, 1 g
Catalog # 8.41379.0001
Supplier:   Merck
3-(2-Hydroxyethyl)indole
Catalog # 8.41379.0001
Supplier:  Merck
CAS Number:  526-55-6
3-(2-Hydroxyethyl)indole

Glass bottle for solids, 1 g

CAS Number:  526-55-6
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol
  • CAS Number:526-55-6
  • Formula:C₁₀H₁₁NO
  • MDL Number:MFCD00005659
  • Molecular Weight:161.2 g/mol
  • Merck Index:12,09930
  • Boiling Pt:358 °C (1013 hPa)
  • Melting Pt:55…57 °C
  • EINECS:208-393-2

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.