Order Entry
Finland
Orders LinkContactUsLinkComponent
Potassium fluoride

Potassium fluoride

Fibre carton, 50 kg
Catalog # 1.05030.9050
Toxic
Supplier:   Merck
Potassium fluoride
Catalog # 1.05030.9050
Supplier:  Merck
CAS Number:  7789-23-3
Potassium fluoride

Fibre carton, 50 kg

CAS Number:  7789-23-3
Price
Inquire For Price
Log in for availability
1 * 50 kg
Product Details & Documents
  • CAS Number:7789-23-3
  • Formula:KF
  • MDL Number:MFCD00011398
  • Molecular Weight:58.1 g/mol
  • Merck Index:13,07714
  • Boiling Pt:1505 °C (1013 hPa)
  • Melting Pt:860 °C
  • Density:2.48 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-151-5
  • UN:1812
  • ADR:6.1,III

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Fibre carton
  • Pk:
    50 kg

Specifications