Order Entry
Finland
Orders LinkContactUsLinkComponent
Vacuum filtration systems, Nalgene®

Vacuum filtration systems, Nalgene®

150 * ml, 47 mm, 0,45 µm
Catalog # 513-1302
   Low Manufacturing Impact
Vacuum filtration systems, Nalgene®
Catalog # 513-1302
   Low Manufacturing Impact
Vacuum filtration systems, Nalgene®
Vacuum filtration systems, Nalgene®

150 * ml, 47 mm, 0,45 µm

Price
159,00 €
Log in for availability
1 * 12 Items
Product Details & Documents

Cellulose nitrate membrane, upper: PP, lower: PS. Ready to use for microbial analysis, water quality or environmental testing.

  • Graduation on upper and lower parts
  • Easy retrieval of the membrane

* with grey grid

Specifications
  • Pore size:
    0,45 µm
  • Capacity:
    150 * ml
  • Membrane Ø:
    47 mm
  • Cat. No.:
    513-1302
  • Environmental certification:
    ISO14001 (Certificate Registration No. EMS 752603)
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact