Order Entry
Finland
Orders LinkContactUsLinkComponent
Objectives for DMW-B1
Plan phase objective (spring)
Catalog # 632-8994
Supplier:   MOTIC
Objectives for DMW-B1
Catalog # 632-8994
Supplier:  MOTIC
Objectives for DMW-B1
Objectives for DMW-B1
Plan phase objective (spring)
Price
231,00 €
Log in for availability
1 * 1 Items
Product Details & Documents
Plan phase objectives for use with the digital microscope DMW-B1.
Specifications
  • Magnification:
    40×/0,65
  • Description:
    Plan phase objective (spring)