Order Entry
Finland
Orders LinkContactUsLinkComponent
Pasteur pipettes

Pasteur pipettes

3 ml, 152,19, 0,50 ml, -, 500 pcs.
Catalog # 612-2815
Supplier:   KARTELL
Pasteur pipettes
Catalog # 612-2815
Supplier:  KARTELL
Pasteur pipettes
Pasteur pipettes

3 ml, 152,19, 0,50 ml, -, 500 pcs.

Price
Inquire For Price
Log in for availability
1 * 500 Items
Product Details & Documents

PE, disposable, sterile or non sterile.

  • Disposable PE pipettes with bulb and graduations
  • The slender tip guarantees precision transfer of the sample
  • Non sterile pipettes are available in multi-packs; sterile pipettes are individually wrapped

 : *Capillary

Specifications
  • Packed:
    500 pcs.
  • Sterile:
    -
  • Capacity:
    3 ml
  • Length:
    152,19
  • Seq:
    6
  • Graduation:
    0,50 ml