Order Entry
Finland
Orders LinkContactUsLinkComponent
Nitromethane 99+% for analysis

Nitromethane 99+% for analysis

Glass bottle, 500 mL
Catalog # ACRO213375000
Flammable
Irritant
Health
Nitromethane 99+% for analysis
Catalog # ACRO213375000
CAS Number:  75-52-5
Nitromethane 99+% for analysis
Nitromethane 99+% for analysis

Glass bottle, 500 mL

CAS Number:  75-52-5
Price
125,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
  • CAS Number:75-52-5
  • Formula:CH₃NO₂
  • MDL Number:MFCD00007400
  • Molecular Weight:61.04 g/mol
  • Merck Index:13,06644
  • Boiling Pt:100…102 °C (760 mmHg)
  • Melting Pt:–29 °C
  • Density:1.12 g/cm³
  • Flash Pt:35 °C
  • Storage Temperature:Ambient
  • EINECS:200-876-6
  • UN:1261
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3805 to 1.3825 (20°C, 589 nm)
  • Water:
    ≤0.5 % (Coulometric)
  • GC:
    ≥99.0 %
  • Residue after evaporation:
    ≤0.005 %
  • Cadmium (Cd):
    ≤0.1 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.5 ppm
  • Lead (Pb):
    ≤0.1 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.