Pyrene 98%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:129-00-0
- Formula:C₁₆H₁₀
- MDL Number:MFCD00004136
- Molecular Weight:202.26 g/mol
- Merck Index:12,08147
- Boiling Pt:393 °C (760 mmHg)
- Melting Pt:148…152 °C
- Density:1,27 g/cm³ (20 °C)
- Flash Pt:210 °C
- Storage Temperature:Ambient
- EINECS:204-927-3
- UN:3077
- ADR:9,III
Documents
ACRO18083
SDS
Specifications
Specification Test Results
- Appearance:Light yellow to yellow to yellow-green Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:147 to 152 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:100 g



