α-Pinene 97% stabilised
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
α-Pinene 97% stabilised
Catalog # ACRO164360050
Supplier: Thermo Fisher Scientific
CAS Number: 80-56-8
Jump to:
Product Details & Documents
- CAS Number:80-56-8
- Formula:C₁₀H₁₆
- MDL Number:MFCD00001339
- Molecular Weight:136.24 g/mol
- Boiling Pt:154…157 °C (760 mmHg)
- Melting Pt:–55 °C
- Density:0.85 g/cm³
- Flash Pt:38 °C
- Storage Temperature:Ambient
- EINECS:219-445-9
- UN:2368
- ADR:3,III
- :Stabilised with α-tocopherol
- :Density: 0.8592 g/cm³ (20 °C)
Documents
ACRO16436
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4640 to 1.4680 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:0.8 % Alpha-tocopherol
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



