4-Hydroxybenzaldehyde 99%
Plastic bottle, 250 g
Supplier: Thermo Fisher Scientific
4-Hydroxybenzaldehyde 99%
Catalog # ACRO162772500
Supplier: Thermo Fisher Scientific
CAS Number: 123-08-0
Jump to:
Product Details & Documents
- CAS Number:123-08-0
- Formula:C₇H₆O₂
- MDL Number:MFCD00006939
- Molecular Weight:122.12 g/mol
- Merck Index:13,04836
- Boiling Pt:310 °C (1013 hPa)
- Melting Pt:114…118 °C
- Density:1,13 g/cm³ (20 °C)
- Flash Pt:174 °C
- Storage Temperature:Ambient
- EINECS:204-599-1
Specifications
Specification Test Results
- Appearance:Yellow to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:114 to 118 °C
- GC:≥98.5 %
- Stabilizer:2 to 5 ppm BHT
- Residue after evaporation:≤0.001 %
- Acidity (CH3COOH):≤0.002
- Peroxides:≤0.005 % (as H2O2)
Specifications
- Pack type:Plastic bottle
- Pk:250 g



