Order Entry
Finland
Orders LinkContactUsLinkComponent
Nitromethane 99%, pure

Nitromethane 99%, pure

Glass bottle, 2,5 L
Catalog # ACRO148510025
Flammable
Irritant
Health
Nitromethane 99%, pure
Catalog # ACRO148510025
CAS Number:  75-52-5
Nitromethane 99%, pure
Nitromethane 99%, pure

Glass bottle, 2,5 L

CAS Number:  75-52-5
Price
336,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
  • CAS Number:75-52-5
  • Formula:CH₃NO₂
  • MDL Number:MFCD00007400
  • Molecular Weight:61.04 g/mol
  • Merck Index:13,06644
  • Boiling Pt:100…102 °C (760 mmHg)
  • Melting Pt:–29 °C
  • Density:1.12 g/cm³
  • Flash Pt:35 °C
  • Storage Temperature:Ambient
  • EINECS:200-876-6
  • UN:1261
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3810 to 1.3830 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤20 APHA

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications