Order Entry
Finland
ContactUsLinkComponent
IFNgamma, Mouse, Biotrak Assay, ELISA - 96 wells (Range: 37-3000pg/ml. Sensitivity: <10pg/ml. Sample volume: 50ul. 1.5L wash buffer supplied. Calibrated to the NIBSC standard Gg02-901-533) 1 * 96 Assays
Catalog # RPN2707
Supplier:  Cytiva
IFNgamma, Mouse, Biotrak Assay, ELISA - 96 wells (Range: 37-3000pg/ml. Sensitivity: <10pg/ml. Sample volume: 50ul. 1.5L wash buffer supplied. Calibrated to the NIBSC standard Gg02-901-533) 1 * 96 Assays
Catalog # RPN2707
Supplier:  Cytiva