Order Entry
Finland
Orders LinkContactUsLinkComponent
Fumaric acid 99+%

Fumaric acid 99+%

Plastic bottle, 2,5 kg
Catalog # ACRO119750025
Irritant
Fumaric acid 99+%
Catalog # ACRO119750025
CAS Number:  110-17-8
Fumaric acid 99+%
Fumaric acid 99+%

Plastic bottle, 2,5 kg

CAS Number:  110-17-8
Price
99,20 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  trans-1,2-Ethylentricarboxylic acid
  • CAS Number:110-17-8
  • Formula:C₄H₄O₄
  • MDL Number:MFCD00002700
  • Molecular Weight:116.07 g/mol
  • Merck Index:13,04308
  • Boiling Pt:356 °C (1013 hPa)
  • Melting Pt:295…300 °C (subl.)
  • Density:1.62 g/cm³
  • Flash Pt:230 °C
  • Storage Temperature:Ambient
  • EINECS:203-743-0

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    295 to 300 °C
  • Water:
    ≤0.25 % (K.F.)
  • HPLC:
    ≥99.0 %
  • Titration with NaOH:
    99.0 to 101.0 % (On anhydrous substance)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.