4'-Fluoroacetophenone 99%
Glass bottle, 100 mL
: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
- :403-42-9
- :C₈H₇FO
- :MFCD00000354
- :138.14 g/mol
- :196 °C (760 mmHg)
- :4 °C
- :1.13 g/cm³
- :71 °C
- :Ambient
- :206-960-9
Documents
ACRO11926
SDS
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5090 to 1.5120 (20°C, 589 nm)
- GC:≥98.5 %
- Pack type:Glass bottle
- Pk:100 mL
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



