4'-Fluoroacetophenone 99%
Glass bottle, 25 mL
Supplier: Thermo Fisher Scientific
4'-Fluoroacetophenone 99%
Catalog # ACRO119260250
Supplier: Thermo Fisher Scientific
CAS Number: 403-42-9
Jump to:
Product Details & Documents
Synonyms: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
- CAS Number:403-42-9
- Formula:C₈H₇FO
- MDL Number:MFCD00000354
- Molecular Weight:138.14 g/mol
- Boiling Pt:196 °C (760 mmHg)
- Melting Pt:4 °C
- Density:1.13 g/cm³
- Flash Pt:71 °C
- Storage Temperature:Ambient
- EINECS:206-960-9
Documents
ACRO11926
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5090 to 1.5120 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:25 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



