2,6-Dichlorophenol 99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
2,6-Dichlorophenol 99%
Catalog # ACRO113590250
Supplier: Thermo Fisher Scientific
CAS Number: 87-65-0
Jump to:
Product Details & Documents
- CAS Number:87-65-0
- Formula:C₆H₄Cl₂O
- MDL Number:MFCD00002176
- Molecular Weight:163 g/mol
- Merck Index:13,03099
- Boiling Pt:∼219 °C (1013 hPa)
- Melting Pt:65…68 °C
- Density:1,653 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:201-761-3
- UN:2020
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:White to yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:65 to 68 °C
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



