2,6-Dichlorophenol 99%
Glass bottle for solids, 1 kg
- :87-65-0
- :C₆H₄Cl₂O
- :MFCD00002176
- :163 g/mol
- :13,03099
- :∼219 °C (1013 hPa)
- :65…68 °C
- :1,653 g/cm³ (20 °C)
- :Ambient
- :201-761-3
- :2020
- :6.1,III
Documents
- Appearance:White to yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:65 to 68 °C
- GC:≥98.5 %
- Pack type:Glass bottle for solids
- Pk:1 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



