Order Entry
Finland
Orders LinkContactUsLinkComponent
Accessories

Accessories

Dummy electrode, PTFE, round tip, max flow rate 100 ml/min
Catalog # 18-1111-03
Supplier:   Cytiva
Accessories
Catalog # 18-1111-03
Supplier:  Cytiva
Accessories
Accessories

Dummy electrode, PTFE, round tip, max flow rate 100 ml/min

Price
186,00 €
Log in for availability
1 * 1 Items
Specifications
  • Description:
    Dummy electrode, PTFE, round tip, max flow rate 100 ml/min
  • Cat. No.:
    18-1111-03
  • W×D×H:
    260×370×100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.