Order Entry
Finland
ContactUsLinkComponent
Bottletop dispenser Dispensette Organic Analog-adjustable, DE-M marking, 2,5-25 ml, with SafetyPrime venting valve 1 * 1 items
Catalog # 613-3339
Supplier:  Brand
Bottletop dispenser Dispensette Organic Analog-adjustable, DE-M marking, 2,5-25 ml, with SafetyPrime venting valve 1 * 1 items
Catalog # 613-3339
Supplier:  Brand