Bellow connectors
19/26 NS, 105 mm, 19/26 NS
BOHLH906-04
Bellow connectors, made of PTFE, with sharp folds and sealing rings on the outside of the cone.
- Universal chemical resistance
- Suitable for vacuum applications
- Temperature resistance from −200 to +250 °C
- Compensates vibrations from vacuum pumps
- Suiltable for length compensation of heated columns and angular misalignment
- Cone size:19/26 NS
- Socket size:19/26 NS
- Seq:02
- Total length:105 mm



