Order Entry
Finland
Orders LinkContactUsLinkComponent
2-Methylindoline 99+%

2-Methylindoline 99+%

Glass bottle, 100 mL
Catalog # ACRO275881000
Irritant
2-Methylindoline 99+%
Catalog # ACRO275881000
CAS Number:  6872-06-6
2-Methylindoline 99+%
2-Methylindoline 99+%

Glass bottle, 100 mL

CAS Number:  6872-06-6
Price
112,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
  • CAS Number:6872-06-6
  • Formula:C₉H₁₁N
  • MDL Number:MFCD00005706
  • Molecular Weight:133.19 g/mol
  • Boiling Pt:227 °C (1013mbar)
  • Melting Pt:–51 °C
  • Density:1.02 g/cm³
  • Flash Pt:103 °C
  • EINECS:229-971-0

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5680 to 1.5700 (20°C, 589 nm)
  • Water:
    ≤0.5 % (K.F.)
  • GC:
    ≥99.0 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications