Order Entry
Finland
Orders LinkContactUsLinkComponent
2-Methylindoline 99+%

2-Methylindoline 99+%

Glass bottle, 5 mL
Catalog # ACRO275880050
Irritant
2-Methylindoline 99+%
Catalog # ACRO275880050
CAS Number:  6872-06-6
2-Methylindoline 99+%
2-Methylindoline 99+%

Glass bottle, 5 mL

CAS Number:  6872-06-6
Price
32,10 €
Log in for availability
1 * 5 mL
Product Details & Documents
  • CAS Number:6872-06-6
  • Formula:C₉H₁₁N
  • MDL Number:MFCD00005706
  • Molecular Weight:133.19 g/mol
  • Boiling Pt:227 °C (1013mbar)
  • Melting Pt:–51 °C
  • Density:1.02 g/cm³
  • Flash Pt:103 °C
  • EINECS:229-971-0

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5680 to 1.5700 (20°C, 589 nm)
  • Water:
    ≤0.5 % (K.F.)
  • GC:
    ≥99.0 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.