Order Entry
Finland
ContactUsLinkComponent
Stirring bar, magnetic, spinvane, triangular, PTFE, fits vials 1ml, size width 5.6mm, height 9.6mm, depth 4.8mm, shaped vanes with c ross-mounted, triangular shape permits rotation in small conical-shaped tubes and micro vials. 1 * 1 items
Catalog # BELAF371340000
Stirring bar, magnetic, spinvane, triangular, PTFE, fits vials 1ml, size width 5.6mm, height 9.6mm, depth 4.8mm, shaped vanes with c ross-mounted, triangular shape permits rotation in small conical-shaped tubes and micro vials. 1 * 1 items
Catalog # BELAF371340000
Frequently Bought Together