Order Entry
Finland
Orders LinkContactUsLinkComponent
Fumaric acid 99%

Fumaric acid 99%

Bottle
Catalog # APOSOR1013408-25KG
Irritant
New Product
Supplier:   Apollo Scientific
Fumaric acid 99%
Catalog # APOSOR1013408-25KG
Supplier:  Apollo Scientific
New Product
CAS Number:  110-17-8
Fumaric acid 99%

Bottle

CAS Number:  110-17-8
Price
2 580,00 €
Log in for availability
1 * 25 kg
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  trans-1,2-Ethylentricarboxylic acid
  • CAS Number:110-17-8
  • Formula:C₄H₄O₄
  • MDL Number:MFCD00002700
  • Molecular Weight:116,07 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.