Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Fumaric acid 99%

Bottle
Catalog # APOSOR10134082.5KG
Irritant
New Product
Supplier:   Apollo Scientific
Fumaric acid 99%
Catalog # APOSOR10134082.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  110-17-8
Fumaric acid 99%

Bottle

CAS Number:  110-17-8
Price
265,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  trans-1,2-Ethylentricarboxylic acid
  • CAS Number:110-17-8
  • Formula:C₄H₄O₄
  • MDL Number:MFCD00002700
  • Molecular Weight:116,07 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications