Order Entry
Finland
Orders LinkContactUsLinkComponent
(1s)-(-)-Alpha-Pinene 98%

(1s)-(-)-Alpha-Pinene 98%

Bottle
Catalog # APOSOR1028057100ML
New Product
Supplier:   Apollo Scientific
(1s)-(-)-Alpha-Pinene 98%
Catalog # APOSOR1028057100ML
Supplier:  Apollo Scientific
New Product
CAS Number:  7785-26-4
(1s)-(-)-Alpha-Pinene 98%

Bottle

CAS Number:  7785-26-4
Price
25,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  (-)-α-Pinene, (1S)-(-)-2-Pinene, (-)-Pin-2(3)-ene, L(-)-α-Pinene
  • CAS Number:7785-26-4
  • Formula:C₁₀H₁₆
  • Molecular Weight:136,23 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Suositukset

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.