Order Entry
Finland
Orders LinkContactUsLinkComponent
Ac4ManNAz ≥98%

Ac4ManNAz ≥98%

Bottle
Catalog # APOSBUP14330-100MG
New Product
Supplier:   Apollo Scientific
Ac4ManNAz ≥98%
Catalog # APOSBUP14330-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  361154-30-5
Ac4ManNAz ≥98%

Bottle

CAS Number:  361154-30-5
Price
514,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Ac4ManNAz
  • CAS Number:361154-30-5
  • Formula:C₁₆H₂₂N₄O₁₀
  • Molecular Weight:430,37 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications