Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Ac4ManNAz ≥98%

Bottle
Catalog # APOSBUP14330-25MG
New Product
Supplier:   Apollo Scientific
Ac4ManNAz ≥98%
Catalog # APOSBUP14330-25MG
Supplier:  Apollo Scientific
New Product
CAS Number:  361154-30-5
Ac4ManNAz ≥98%

Bottle

CAS Number:  361154-30-5
Price
231,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Ac4ManNAz
  • CAS Number:361154-30-5
  • Formula:C₁₆H₂₂N₄O₁₀
  • Molecular Weight:430,37 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.