Order Entry
Finland
Orders LinkContactUsLinkComponent
L(-)-Mannose

L(-)-Mannose

Bottle
Catalog # APOSBIFS0112-500MG
Irritant
New Product
Supplier:   Apollo Scientific
L(-)-Mannose
Catalog # APOSBIFS0112-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  10030-80-5
L(-)-Mannose

Bottle

CAS Number:  10030-80-5
Price
133,00 €
Log in for availability
1 * 500 mg
Product Details & Documents
Synonyms:  (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose
  • CAS Number:10030-80-5
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00136021
  • Molecular Weight:180.16 g/mol
  • Melting Pt:129…131 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.