Order Entry
Finland
Orders LinkContactUsLinkComponent
Mouse recombinant Acrp30

Mouse recombinant Acrp30

2 µg
Catalog # APOSBITP1001-2UG
New Product
Supplier:   Apollo Scientific
Mouse recombinant Acrp30
Catalog # APOSBITP1001-2UG
Supplier:  Apollo Scientific
New Product
CAS Number:  
Mouse recombinant Acrp30

2 µg

CAS Number:  
Price
162,00 €
Log in for availability
1 * 1 Each
Product Details & Documents
Specifications

Specifications

  • Pk:
    2 µg
  • Cat. No.:
    APOSBITP1001-2UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Acrp30
  • Protein synonyms:
    Adiponectin
  • Species:
    Mouse

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.