Cholesterol 95%
Bottle
Catalog # APOSOR1030208-25G
New Product
Supplier: Apollo Scientific
Cholesterol 95%
Catalog # APOSOR1030208-25G
Supplier: Apollo Scientific
New Product
CAS Number: 57-88-5
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
- CAS Number:57-88-5
- Formula:C₂₇H₄₆O
- MDL Number:MFCD00003646
- Molecular Weight:386,55 g/mol
Specifications
Specifications
- Pack type:Bottle



