Order Entry
Finland
Orders LinkContactUsLinkComponent
2,6-Dichlorophenol 98%

2,6-Dichlorophenol 98%

Bottle
Catalog # APOSOR0357-1KG
Corrosive
Irritant
Environment
New Product
Supplier:   Apollo Scientific
2,6-Dichlorophenol 98%
Catalog # APOSOR0357-1KG
Supplier:  Apollo Scientific
New Product
CAS Number:  87-65-0
2,6-Dichlorophenol 98%

Bottle

CAS Number:  87-65-0
Price
474,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:87-65-0
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002176
  • Molecular Weight:163 g/mol
  • Boiling Pt:218…220 °C
  • Melting Pt:64…68 °C
  • Storage Temperature:Ambient
  • EINECS:201-761-3
  • UN:2020
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications