2,6-Dichlorophenol 98%
Bottle
Catalog # APOSOR0357-250G
New Product
Supplier: Apollo Scientific
2,6-Dichlorophenol 98%
Catalog # APOSOR0357-250G
Supplier: Apollo Scientific
New Product
CAS Number: 87-65-0
Siirry kohtaan:
Product Details & Documents
- CAS numero:87-65-0
- Kaava:C₆H₄Cl₂O
- MDL-numero:MFCD00002176
- Molecular Weight:163 g/mol
- Kiehumispiste:218…220 °C
- Sulamispiste:64…68 °C
- Varastointilämpötila:Ambient
- EINECS:201-761-3
- UN:2020
- ADR:6.1,III
Tekniset tiedot
Tekniset tiedot
- Pack type:Bottle
Tekniset tiedot
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



