Xanthydrol
Bottle
Catalog # APOSBIX1001-100G
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 9-Hydroxyxanthene, 9-Xanthenol, 9H-Xanthen-9-ol
- CAS Number:90-46-0
- Formula:C₁₃H₁₀O₂
- MDL Number:MFCD00005057
- Molecular Weight:198.22 g/mol
- Melting Pt:124…127 °C
- Storage Temperature:Refrigerator
Specifications
Specifications
- Pack type:Bottle



