Order Entry
Finland
Orders LinkContactUsLinkComponent
Xanthydrol

Xanthydrol

Bottle
Catalog # APOSBIX1001-1G
New Product
Supplier:   Apollo Scientific
Xanthydrol
Catalog # APOSBIX1001-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  90-46-0
Xanthydrol

Bottle

CAS Number:  90-46-0
Price
22,10 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  9-Hydroxyxanthene, 9-Xanthenol, 9H-Xanthen-9-ol
  • CAS Number:90-46-0
  • Formula:C₁₃H₁₀O₂
  • MDL Number:MFCD00005057
  • Molecular Weight:198.22 g/mol
  • Melting Pt:124…127 °C
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle

Specifications