Order Entry
Finland
Orders LinkContactUsLinkComponent
Methabenzthiazuron

Methabenzthiazuron

Methabenzthiazuron
Catalog # EHERC14960000
Supplier:   EHRENSTORFER
Methabenzthiazuron
Catalog # EHERC14960000
Supplier:  EHRENSTORFER
CAS Number:  
Methabenzthiazuron
Methabenzthiazuron

Methabenzthiazuron

CAS Number:  
Price
63,70 €
Log in for availability
1 * 0,1 g
Product Details & Documents
  • SMILES: CNC(=O)N(C)c1nc2ccccc2s1
  • Inchi: InChI=1S/C10H11N3OS/c1-11-9(14)13(2)10-12-7-5-3-4-6-8(7)15-10/h3-6H,1-2H3,(H,11,14)
  • IUPAC: 1-(1,3-benzothiazol-2-yl)-1,3-dimethylurea
  • Packing group: III
Specifications

Specifications

  • Description:
    Methabenzthiazuron
  • UN No.:
    3077
  • Cat. No.:
    EHERC14960000
  • Formula:
    C₁₀H₁₁N₃OS
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS07, GHS09
  • GHS signal:
    Warning
  • Synonym(s):
    Urea, 1-(2-benzothiazolyl)-1,3-dimethyl- (8CI);N-2-Benzothiazolyl-N,N'-dimethylurea;1,3-Dimethyl-3-(2-benzothiazolyl)urea;1-(2-Benzothiazolyl)-1,3-dimethylurea;5633;BAY 5653;Bayer 5653;Bayer 74283;Metabenzthiazuron;Methabenzthiazuron;Methbenzthiazuron;Methibenzuron;Preparation 5633;Tribunil
  • Molecular weight:
    221,28
  • CAS No.:
    18691-97-9
  • Shipping conditions:
    RT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.