Fenuron
Fenuron
Catalog # EHERC13620000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CN(C)C(=O)Nc1ccccc1
- Inchi: InChI=1S/C9H12N2O/c1-11(2)9(12)10-8-6-4-3-5-7-8/h3-7H,1-2H3,(H,10,12)
- IUPAC: 1,1-dimethyl-3-phenylurea
- Packing group: III
Specifications
Specifications
- Description:Fenuron
- UN No.:3077
- Cat. No.:EHERC13620000
- Formula:C₉H₁₂N₂O
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Urea, N,N-dimethyl-N'-phenyl-;Urea, 1,1-dimethyl-3-phenyl- (8CI);N,N-Dimethyl-N'-phenylurea;1,1-Dimethyl-3-phenylurea;1-Phenyl-3,3-dimethylurea;3-Phenyl-1,1-dimethylurea;Amicure UR;Dibar;Dybar;Dyhard 300;Dyhard RU 300;Dyhard UR 300;Falisilvan;Fenuron;Fikure 62U;N-Phenyl-N',N'-dimethylurea;Omicure 94;Omicure U 405;PUD;UR 300
- Molecular weight:164,2
- CAS No.:101-42-8
- Shipping conditions:RT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



