DNOC
DNOC
Catalog # EHERC13050000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Cc1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-] Inchi: InChI=1S/C7H6N2O5/c1-4-2-5(8(11)12)3-6(7(4)10)9(13)14/h2-3,10H,1H3 IUPAC: 2-methyl-4,6-dinitrophenol
- Packing group: II
Specifications
Specifications
- Description:DNOC
- UN No.:1598
- Cat. No.:EHERC13050000
- Formula:C₇H₆N₂O₅
- Storage conditions:20 ℃
- Hazard class description:GHS05, GHS06, GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Phenol, 2-methyl-4,6-dinitro-; o-Cresol, 4,6-dinitro- (8CI); 2-Methyl-4,6-dinitrophenol; 2,4-Dinitro-6-methylphenol; 3,5-Dinitro-2-hydroxytoluene; 4,6-Dinitro-2-methylphenol; 4,6-Dinitro-o-cresol; 6-Methyl-2,4-dinitrophenol; Arborol; DNOC; Degrassan; Dekrysil; Detal; Dillex; Dinitro; Dinitro-o-cresol; Dinitrocresol; Dinitrodendtroxal; Dinitrol; Dinoc; Dinurania; Ditrosol; Effusan; Effusan 3436; Elgetol; Elgetol 30; Elipol; Extrar; Flavin-Sandoz; Hedolit; Hedolite; K III; K IV; Kreozan; Krezotol 50; Lipan; NSC 2082; Neudorff DN 50; Nitrofan; Prokarbol; Rafex; Rafex 35; Raphatox; Sandolin; Sandolin A; Selinon; Sinox; Winterwash
- Molecular weight:198,13
- CAS No.:534-52-1
- Shipping conditions:RT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



