Monolinuron
Monolinuron
Catalog # EHERC15310000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CON(C)C(=O)Nc1ccc(Cl)cc1
- Inchi: InChI=1S/C9H11ClN2O2/c1-12(14-2)9(13)11-8-5-3-7(10)4-6-8/h3-6H,1-2H3,(H,11,13)
- IUPAC: 3-(4-chlorophenyl)-1-methoxy-1-methylurea
- Packing group: III
Specifications
Specifications
- Description:Monolinuron
- UN No.:3077
- Cat. No.:EHERC15310000
- Formula:C₉H₁₁ClN₂O₂
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Warning
- Molecular weight:214,65
- CAS No.:1746-81-2
- Shipping conditions:RT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



