Fenthion
Fenthion
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COP(=S)(OC)Oc1ccc(SC)c(C)c1
- Inchi: InChI=1S/C10H15O3PS2/c1-8-7-9(5-6-10(8)16-4)13-14(15,11-2)12-3/h5-7H,1-4H3
- IUPAC: dimethoxy-(3-methyl-4-methylsulfanylphenoxy)-sulfanylidene-λ^{5}-phosphane
- Packing group: III
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:Fenthion
- Cat. No.:EHERC13580000
- UN No.:3018
- Formula:C₁₀H₁₅O₃PS₂
- Storage conditions:4 °C
- Hazard class description:GHS06, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Fenthion;Phosphorothioic Acid O,O-Dimethyl O-[3-Methyl-4-(methylthio)phenyl] Ester;ENT-25540;Baycid;Bayer 29493;Mercaptophos;O,O-Dimethyl O-[4-(Methylthio)-m-tolyl] thiophosphate;
- Molecular weight:278,33
- CAS No.:55-38-9
- Shipping conditions:RT



