Order Entry
Finland
Orders LinkContactUsLinkComponent
Prothioconazole

Prothioconazole

Prothioconazole
Catalog # EHERC16555000
Supplier:   EHRENSTORFER
Prothioconazole
Catalog # EHERC16555000
Supplier:  EHRENSTORFER
CAS Number:  
Prothioconazole
Prothioconazole

Prothioconazole

CAS Number:  
Price
346,00 €
Log in for availability
1 * 0,1 g
Product Details & Documents
  • SMILES: OC(CN1NC=NC1=S)(Cc2ccccc2Cl)C3(Cl)CC3
  • Inchi: InChI=1S/C14H15Cl2N3OS/c15-11-4-2-1-3-10(11)7-14(20,13(16)5-6-13)8-19-12(21)17-9-18-19/h1-4,9,20H,5-8H2,(H,17,18,21)
  • IUPAC: 2-[2-(1-chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-1H-1,2,4-triazole-3-thione
  • Packing group: III
Specifications

Specifications

  • Description:
    Prothioconazole
  • UN No.:
    3077
  • Cat. No.:
    EHERC16555000
  • Formula:
    C₁₄H₁₅Cl₂N₃OS
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS08, GHS09
  • GHS signal:
    Danger
  • Synonym(s):
    2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-2,4-dihydro-3H-1,2,4-triazole-3-thione; JAU 6476; Joao; Proline (fungicide); Proline 275; Proline 480SC; Prothioconazole; Redigo; Rudis
  • Molecular weight:
    344,26
  • CAS No.:
    178928-70-6
  • Shipping conditions:
    RT

Specifications