Order Entry
Finland
Orders LinkContactUsLinkComponent
Phloretin ≥98%

Phloretin ≥98%

New Product
Supplier:   Apollo Scientific
Phloretin ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Phloretin ≥98%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

  • CAS Number:60-82-2
  • Formula:C₁₅H₁₄O₅
  • Molecular Weight:274,27 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 75,00 €
    75,00 €
    1 * 500 mg
       75,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 331,00 €
    331,00 €
    1 * 10 g
       331,00 €

    Specifications:
    New Product
    New Product