Order Entry
Finland
Orders LinkContactUsLinkComponent
(1s)-(-)-Alpha-Pinene 98%

(1s)-(-)-Alpha-Pinene 98%

New Product
Supplier:   Apollo Scientific
(1s)-(-)-Alpha-Pinene 98%
Supplier:  Apollo Scientific
New Product
Certificates
(1s)-(-)-Alpha-Pinene 98%

4 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  (-)-α-Pinene, (1S)-(-)-2-Pinene, (-)-Pin-2(3)-ene, L(-)-α-Pinene
  • CAS Number:7785-26-4
  • Formula:C₁₀H₁₆
  • Molecular Weight:136,23 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 22,10 €
    22,10 €
    1 * 25 mL
       22,10 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 25,00 €
    25,00 €
    1 * 100 mL
       25,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 41,20 €
    41,20 €
    1 * 500 mL
       41,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 200,00 €
    200,00 €
    1 * 2,5 L
       200,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.